Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:ABRO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:23172
- Antigen Synonyms:Protein FAM175B|Em:AC068896.4|KIAA0157BRISC complex subunit Abro1|Abraxas brother protein 1|ABRO1abraxas brother 1|family with sequence similarity 175|member B|FLJ22338
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 390 and 415 of human abraxas brother 1 using the numbering given in entry NP_115558.3 (GeneID 23172).
- Purification:Immunogen affinity purified
- Cat. No.:102110-974
- Supplier no.:NB100-60449
The ABRO Antibody from Novus Biologicals is a rabbit polyclonal antibody to ABRO. This antibody reacts with human. The ABRO Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: ABRO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human