Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:5-Hydroxytryptamine receptor 4
- Antigen Symbol:5-HT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Size:0.05 ml
- Gene ID:3360
- Antigen Synonyms:5-HT4R|5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor|cardiac 5-HT4 receptor|Serotonin receptor 4|5-HT-4
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human 5HT4 Receptor.
- Purification:Immunogen affinity purified
- Cat. No.:102253-430
- Supplier no.:NLS655
The 5-HT4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to 5-HT4. This antibody reacts with human, monkey. The 5-HT4 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: 5-HT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey