Order Entry
Puerto Rico
ContactUsLinkComponent
pre-mRNA cleavage factor I (59 kDa subunit) Lysate (Adult Normal), Novus Biologicals (NBP2-11084)
pre-mRNA cleavage factor I (59 kDa subunit) Lysate (Adult Normal), Novus Biologicals (NBP2-11084)
Catalog # 102248-954
Supplier:  Novus Biologicals
CAS Number:  
undefined
pre-mRNA cleavage factor I (59 kDa subunit) Lysate (Adult Normal), Novus Biologicals (NBP2-11084)
Catalog # 102248-954
Supplier:  Novus Biologicals
Supplier Number:  NBP2-11084
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cell Format:
    Cryopreserved
  • Size:
    0.1 mg
  • Tested Applications:
    WB
  • Cat. No.:
    102248-954
  • Supplier no.:
    NBP2-11084

Specifications

About this item

pre-mRNA cleavage factor I (59 kDa subunit) Lysate (Adult Normal)

Tested Applications: Western Blot.