To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:TIP41 toR signaling pathway Regulator-like
- Antigen Symbol:TIPRL
- Clonality:Monoclonal
- Clone:3D11
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:261726
- Antigen Synonyms:Putative MAPK-activating protein PM10|dJ69E11.3|TIP41Type 2A-interacting protein|TIP41|TOR signalling pathway regulator-like (S. cerevisiae)|MGC3794|putative MAPK activating protein|TIP41-like protein|TIPTIP41|TOR signaling pathway regulator-like (S. cerevisiae)
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human TIPRL (NP_690866) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102231-274
- Supplier no.:NBP2-02269
The TIPRL Antibody (3D11) from Novus Biologicals is a mouse monoclonal antibody to TIPRL. This antibody reacts with human. The TIPRL Antibody (3D11) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TIPRL
Clonality: Monoclonal
Clone: 3D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human