Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome (prosome macropain) 26S Subunit non-ATPase 11
- Antigen Symbol:PSMD11
- Clonality:Monoclonal
- Clone:1F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:C. elegans,Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:5717
- Antigen Synonyms:S9,26S proteasome non-ATPase regulatory subunit 11|non-ATPase|MGC3844,26S proteasome regulatory subunit RPN6|proteasome (prosome|macropain) 26S subunit|11,26S proteasome regulatory subunit p44.5|p44.5,26S proteasome regulatory subunit 9|Rpn6,26S proteasome regulatory subunit S9
- Storage Buffer:PBS (pH 7.4) and 10% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Recombinant human PSMD11 (1-422aa) purified from E. coli (NP_002806).
- Purification:Protein G purified
- Cat. No.:102163-256
- Supplier no.:NBP1-30252
Specifications
About this item
The PSMD11 Antibody (1F4) from Novus Biologicals is a mouse monoclonal antibody to PSMD11. This antibody reacts with human, mouse, c. elegans. The PSMD11 Antibody (1F4) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: PSMD11
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, C. elegans