Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Nitrotyrosine
- Antigen Symbol:Nitro-Tyr
- Clonality:Monoclonal
- Clone:11C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Multiple
- Western Blot:Yes
- Size:1 ml
- Form:Lyophilized
- Antigen Synonyms:Nitrotyrosine
- Storage Buffer:Cell culture supernatant
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:KLH-Nitrotyrosine
- Purification:Unpurified
- Cat. No.:102110-372
- Supplier no.:NB100-57383
Specifications
About this item
The Nitrotyrosine Antibody (11C2) from Novus Biologicals is a mouse monoclonal antibody to Nitrotyrosine. This antibody reacts with all species. The Nitrotyrosine Antibody (11C2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: 11C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: All Species