Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Lymphotactin
- Antigen Symbol:XCL1
- Clonality:Monoclonal
- Clone:MM0604-4G24
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Form:Lyophilized
- Gene ID:6375
- Antigen Synonyms:SCM-1|SCM-1a|small inducible cytokine subfamily C|single cysteine motif 1a|ATACSmall-inducible cytokine C1|Cytokine SCM-1|SCYC1SCM1|LPTNSCM-1-alpha|chemokine (C motif) ligand 1|lymphotaxin|C motif chemokine 1|lymphotactin|LTNXC chemokine ligand 1|member 1 (lymphotactin)|SCM1A
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Human Lymphotactin
- Purification:Protein G purified
- Cat. No.:102250-744
- Supplier no.:NBP2-12004
Specifications
About this item
The XCL1 / Lymphotactin Antibody (MM0604-4G24) from Novus Biologicals is a mouse monoclonal antibody to XCL1 / Lymphotactin. This antibody reacts with human. The XCL1 / Lymphotactin Antibody (MM0604-4G24) has been validated for the following applications: Western Blot.
Type: Primary
Antigen: XCL1
Clonality: Monoclonal
Clone: MM0604-4G24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human