Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:KSHV ORF26
- Clonality:Monoclonal
- Clone:2F6B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Antigen Synonyms:ORF45|Human Herpes Virus 8|KSHV|Kaposi's sarcoma herpes virus|HHV8
- Storage Buffer:Ascites
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Purified recombinant fragment of KSHV ORF26 expressed in E. Coli.
- Purification:Unpurified
- Cat. No.:102177-088
- Supplier no.:NBP1-47357
Specifications
About this item
Mouse Monoclonal KSHV ORF26 Antibody (2F6B8). Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.
Type: Primary
Antigen: KSHV ORF26
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human