Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Kir2.1
- Clonality:Monoclonal
- Clone:S112B-14
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:3759
- Antigen Synonyms:IRK1LQT7|subfamily J|Kir2.1|IRK-1|inward rectifier K+ channel KIR2.1|Inward rectifier K(+) channel Kir2.1|hIRK1|Potassium channel|member 2|SQT3|inwardly rectifying subfamily J member 2|Cardiac inward rectifier potassium channel|HHBIRK1|inward rectifier potassium channel 2|HHIRK1|potassium inwardly-rectifying channel
- Storage Buffer:PBS (pH 7.4) and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Fusion protein amino acids 41-64 and 189-428 of mouse Kir2.1
- Purification:Protein G purified
- Cat. No.:102252-596
- Supplier no.:NBP2-12900
Specifications
About this item
Mouse Monoclonal Kir2.1 Antibody (S112B-14). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.
Recommended Dilutions: Immunohistochemistry (Parafin): 0.1-1.0 ?g/mL
Type: Primary
Antigen: KIR2.1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human