Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:IgM F(c) fragment
- Antigen Symbol:IgM F(c)
- Clonality:Monoclonal
- Clone:CH2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C in the dark.
- Concentration:1 mg/ml
- Immunogen:Human IgM.
- Purification:Immunogen affinity purified
- Cat. No.:102174-394
- Supplier no.:NBP1-44935
Specifications
About this item
Mouse Monoclonal IgM Antibody (CH2) [FITC]. Tested Applications: Western Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.
Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human