To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Eotaxin 2
- Antigen Symbol:CCL24/Eotaxin-2/MPIF-2
- Clonality:Monoclonal
- Clone:61016.11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:6369
- Antigen Synonyms:SCYA24CK-beta-6|Ckb-6|Eotaxin-2|member 24|MPIF-2Small-inducible cytokine A24|chemokine (C-C motif) ligand 24|Eosinophil chemotactic protein 2|MPIF2Myeloid progenitor inhibitory factor 2
- Storage Buffer:PBS
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Purified recombinant human eotaxin 2, expressed in E. coli as immunogen.
- Purification:Protein G purified
- Cat. No.:102126-652
- Supplier no.:NB120-10370
Specifications
About this item
The CCL24 / Eotaxin-2 / MPIF-2 Antibody (61016.11) from Novus Biologicals is a mouse monoclonal antibody to CCL24 / Eotaxin-2 / MPIF-2. This antibody reacts with human. The CCL24 / Eotaxin-2 / MPIF-2 Antibody (61016.11) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CCL24/Eotaxin-2/MPIF-2
Clonality: Monoclonal
Clone: 61016.11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human