Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Drebrin 1
- Clonality:Monoclonal
- Clone:M2F6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Chicken,Human,Bovine,Rat,Mouse,Cat
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:1627
- Antigen Synonyms:drebrin-1|D0S117E|drebrin E2|drebrin E|drebrin 1|DKFZp434D064|Developmentally-regulated brain protein|drebrin
- Storage Buffer:PBS (pH 7.2) and 50% Glycerol
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Chicken Drebrin E protein.
- Purification:Protein A purified
- Cat. No.:102106-430
- Supplier no.:NB100-1951
Specifications
About this item
The Drebrin 1 Antibody (M2F6) from Novus Biologicals is a mouse monoclonal antibody to Drebrin 1. This antibody reacts with human, mouse, rat, bovine, chicken, feline, rabbit. The Drebrin 1 Antibody (M2F6) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml
Type: Primary
Antigen: Drebrin 1
Clonality: Monoclonal
Clone: M2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Chicken, Feline, Rabbit