The DMC1 Antibody (1D12 / 4) from Novus Biologicals is a mouse monoclonal antibody to DMC1. This antibody reacts with human, mouse. The DMC1 Antibody (1D12 / 4) has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: DMC1
Clonality: Monoclonal
Clone: 1D12/4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 102106-076
- 102106-056
- Antigen Symbol:
- DMC1
- DMC1
- Antigen Name:
- DNA Meiotic Recombinase 1
- DNA Meiotic Recombinase 1
- Antigen Synonyms:
- HsLim15|DMC1 dosage suppressor of mck1 homolog|meiotic recombination protein DMC1/LIM15 homolog|LIM15yeast homolog) meiosis-specific homologousrecombination|MGC150472|MGC150473|yeast|DMC1 homologue|meiosis-specific homologousrecombination (yeast)|homolog of|disrupted meiotic cDNA1
- HsLim15|DMC1 dosage suppressor of mck1 homolog|meiotic recombination protein DMC1/LIM15 homolog|LIM15yeast homolog) meiosis-specific homologousrecombination|MGC150472|MGC150473|yeast|DMC1 homologue|meiosis-specific homologousrecombination (yeast)|homolog of|disrupted meiotic cDNA1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1D12/4
- 1D12/4
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 11144
- 11144
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- His-tagged human DMC1, overexpressed in E. coli [UniProt# Q14565]
- His-tagged human DMC1, overexpressed in E. coli [UniProt# Q14565]
- Purification:
- Unpurified
- Unpurified
- Storage Buffer:
- Ascites
- Ascites
- Storage Temperature:
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.