Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Cyclic AMP
- Antigen Symbol:Cyclic AMP
- Clonality:Monoclonal
- Clone:9H4C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Size:100 μl
- Antigen Synonyms:Cyclic AMP|3' 5' cyclic adenosine monophosphate|Cyclic adenosine monophosphate
- Storage Buffer:Ascites
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide of cAMP, conjugated to KLH.
- Purification:Unpurified
- Cat. No.:102181-808
- Supplier no.:NBP1-51543
The Cyclic AMP Antibody (9H4C4) from Novus Biologicals is a mouse monoclonal antibody to Cyclic AMP. This antibody reacts with non-species specific. The Cyclic AMP Antibody (9H4C4) has been validated for the following applications: ELISA.
Type: Primary
Antigen: Cyclic AMP
Clonality: Monoclonal
Clone: 9H4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Non-species specific