Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:C1Q And Tumor Necrosis Factor Related Protein 7
- Antigen Symbol:CTRP7
- Clonality:Monoclonal
- Clone:Anna-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:114905
- Antigen Synonyms:C1q and tumor necrosis factor related protein 7|ZACRP7|CTRP7complement C1q tumor necrosis factor-related protein 7|complement-c1q tumor necrosis factor-related protein 7
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Recombinant mouse CTRP7 (C1q tumor necrosis factor-alpha-related protein 7) (aa 18-290).
- Purification:Protein G purified
- Cat. No.:102222-000
- Supplier no.:NBP1-97528
Specifications
About this item
The CTRP7 Antibody (Anna-1) from Novus Biologicals is a mouse monoclonal antibody to CTRP7. This antibody reacts with human, mouse. The CTRP7 Antibody (Anna-1) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CTRP7
Clonality: Monoclonal
Clone: Anna-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse