Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Killer Cell Lectin-like Receptor Subfamily D, Member 1
- Antigen Symbol:CD94
- Clonality:Monoclonal
- Clone:MM0181-5F26
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:3824
- Antigen Synonyms:CD94 antigen|KP43|NK cell receptor|Killer cell lectin-like receptor subfamily D member 1|member 1|killer cell lectin-like receptor subfamily D|CD94natural killer cells antigen CD94
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at -20 to -70C. Avoid freeze-thaw cycles.
- Immunogen:Human CD94
- Purification:Protein G purified
- Cat. No.:102251-106
- Supplier no.:NBP2-12185
Specifications
About this item
The CD94 Antibody (MM0181-5F26) from Novus Biologicals is a mouse monoclonal antibody to CD94. This antibody reacts with human. The CD94 Antibody (MM0181-5F26) has been validated for the following applications: Flow Cytometry, CyTOF-ready.
Type: Primary
Antigen: CD94
Clonality: Monoclonal
Clone: MM0181-5F26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human