Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD9 molecule
- Antigen Symbol:CD9
- Clonality:Monoclonal
- Clone:MM0180-5H15
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Form:Lyophilized
- Gene ID:928
- Antigen Synonyms:P24|TSPAN-29|Leukocyte antigen MIC3|Motility-related protein|motility related protein-1,5H9 antigen|Cell growth-inhibiting gene 2 protein|TSPAN29MIC3CD9 antigen (p24)|MRP-1FLJ99568|DRAP-27|Tetraspanin-29|BA-2/p24 antigen|BA2|CD9 antigen|BTCC-1|CD9 molecule
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Human CD9
- Purification:Protein G purified
- Cat. No.:102251-104
- Supplier no.:NBP2-12184
Specifications
About this item
The CD9 Antibody (MM0180-5H15) from Novus Biologicals is a mouse monoclonal antibody to CD9. This antibody reacts with human. The CD9 Antibody (MM0180-5H15) has been validated for the following applications: Flow Cytometry, CyTOF-ready.
Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: MM0180-5H15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human