Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:CAMP/LL37/FALL39/Cathelicidin
- Antigen Symbol:CAMP
- Clonality:Monoclonal
- Clone:OSX12
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:820
- Antigen Synonyms:FALL-39|LL37|CAP18CRAMP|18 kDa cationic antimicrobial protein|hCAP-18|FALL39CAP-18|cathelicidin antimicrobial peptide
- Storage Buffer:No buffer
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:A synthetic peptide from human Cathelicidin antimicrobial peptide (CAP-18, hCAP-18, antibacterial protein LL-37, CAMP, CRAMP, FALL39) conjugated to an immunogenic carrier protein has been used as the immunogen.
- Purification:Protein A or G purified
- Cat. No.:102176-498
- Supplier no.:NBP1-46781
The CAMP / LL37 / FALL39 / Cathelicidin Antibody (OSX12) from Novus Biologicals is a mouse monoclonal antibody to CAMP / LL37 / FALL39 / Cathelicidin. This antibody reacts with human. The CAMP / LL37 / FALL39 / Cathelicidin Antibody (OSX12) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 5-10 ?g/ml
Type: Primary
Antigen: CAMP
Clonality: Monoclonal
Clone: OSX12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human