To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Complement Component C5a R1
- Antigen Symbol:C5a R1
- Clonality:Monoclonal
- Clone:S5/1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:728
- Antigen Synonyms:C5ARC5a anaphylatoxin receptor|C5a anaphylatoxin chemotactic receptor|C5a-R|complement component 5 receptor 1 (C5a ligand)|complement component 5a receptor 1|C5aR|CD88|C5A|C5R1C5a ligand|CD88 antigen|complement component 5 receptor 1
- Storage Buffer:0.2 um filtered solution in PBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.1 mg/ml
- Immunogen:Synthetic peptide comprising the N-terminal domain of the C5aR (Met1-Asn31)
- Purification:Protein G purified
- Cat. No.:102126-928
- Supplier no.:NB120-11867
Specifications
About this item
The Complement Component C5a R1 Antibody (S5 / 1) from Novus Biologicals is a mouse monoclonal antibody to Complement Component C5a R1. This antibody reacts with human. The Complement Component C5a R1 Antibody (S5 / 1) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Functional.
Type: Primary
Antigen: C5a R1
Clonality: Monoclonal
Clone: S5/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human