Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-BIN3 Mouse Monoclonal Antibody [clone: 8E9]
Anti-BIN3 Mouse Monoclonal Antibody [clone: 8E9]
Catalog # 102233-514
Supplier:  Novus Biologicals
Anti-BIN3 Mouse Monoclonal Antibody [clone: 8E9]
Catalog # 102233-514
Supplier:  Novus Biologicals
Supplier Number:  NBP2-03389
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Bridging integrator 3
  • Antigen Symbol:
    BIN3
  • Clonality:
    Monoclonal
  • Clone:
    8E9
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Primates,
    Human,
    Dog
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    55909
  • Antigen Synonyms:
    MGC14978|bridging integrator 3
  • Storage Buffer:
    PBS (pH 7.3), 1.0% BSA and 50% Glycerol
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    Full length human recombinant protein of human BIN3 (NP_061158) produced in HEK293T cell.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102233-514
  • Supplier no.:
    NBP2-03389

Specifications

About this item

The BIN3 Antibody (8E9) from Novus Biologicals is a mouse monoclonal antibody to BIN3. This antibody reacts with human, canine, primate. The BIN3 Antibody (8E9) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: BIN3
Clonality: Monoclonal
Clone: 8E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Canine, Primate