Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:ATP Binding Cassette, sub-family A, member 1
- Antigen Symbol:ABCA1
- Clonality:Monoclonal
- Clone:HJ1
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Rabbit,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:19
- Antigen Synonyms:sub-family A (ABC1)|ABC-1|MGC165011|TGDATP-binding cassette sub-family A member 1|EC 3.6.3.41|MGC164864|ATP-binding cassette transporter 1|ABC1ATP-binding cassette transporter A1|ATP-binding cassette 1|CERP|HDLDT1|Cholesterol efflux regulatory protein|EC 3.6.3|FLJ14958|EC 2.7.7.8|member 1|membrane-bound|ATP-binding cassette|EC 2.8.1.8
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:50 kDa N-terminal extracellular loop of mouse ABCA1 [UniProt# P41233]
- Purification:Protein G purified
- Cat. No.:102106-594
- Supplier no.:NB100-2068H
Specifications
About this item
The ABCA1 Antibody (HJ1) [HRP] from Novus Biologicals is a mouse monoclonal antibody to ABCA1. This antibody reacts with human, mouse, rat, rabbit. The ABCA1 Antibody (HJ1) [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ABCA1
Clonality: Monoclonal
Clone: HJ1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat, Rabbit