Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant HspBP1 (from E. coli)
Human Recombinant HspBP1 (from E. coli)
Catalog # 102174-062
Supplier:  Novus Biologicals
CAS Number:  
Human Recombinant HspBP1 (from E. coli)
Catalog # 102174-062
Supplier:  Novus Biologicals
Supplier Number:  NBP1-44481
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage Conditions:
    Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
  • Gene ID:
    23640
  • Protein Synonyms:
    hsp70-binding protein 1|Hsp70-binding protein 2|Hsp70-interacting protein 1|hsp70 interacting protein|HSPA (heat shock 70kDa) binding protein|HSPBP|FES1|Hsp70-interacting protein 2|hspBP2|cytoplasmic cochaperone 1|Heat shock protein-binding protein 1|Hsp70 binding protein 1|HspBP1
  • Protein/Peptide Name:
    HspBP1
  • Purity:
    >95% pure by SDS-PAGE
  • Concentration:
    0.5 mg/ml
  • Tested Applications:
    ELISA
  • Cat. No.:
    102174-062
  • Supplier no.:
    NBP1-44481

Specifications

About this item

The Recombinant Human HspBP1 Protein from Novus Biologicals is derived from E. coli. The Recombinant Human HspBP1 Protein has been validated for the following applications: SDS-Page.