Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:SH3PX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:51429
- Antigen Synonyms:sorting nexin-9|MSTP155|MST155|SH3 and PX domain-containing protein 1|SH3PX1WISP|SH3PXD3ASH3 and PX domain-containing protein SH3PX1|WiskSH3 and PX domain-containing protein 3A|SDP1|Protein SDP1|sorting nexin 9
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-HKGAIEKVKESDK corresponding to internal region according to NP_057308.1.
- Tested Applications:, Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102108-186
- Supplier no.:NB100-2813
The SH3PX1 Antibody from Novus Biologicals is a goat polyclonal antibody to SH3PX1. This antibody reacts with human. The SH3PX1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Peptide ELISA.
Type: Primary
Antigen: SH3PX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human