Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:POU Class 3 Homeobox 3
- Antigen Symbol:POU3F3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Epitope:HMLSHAHQWVTAL - internal region
- Gene ID:5455
- Antigen Synonyms:POU domain class 3|OTF-8|Octamer-binding protein 8|BRN1brn-1|class 3|Octamer-binding transcription factor 8|Brain-specific homeobox/POU domain protein 1|OTF8oct-8|transcription factor 3|brain-1|POU domain|Brn-1|POU class 3 homeobox 3|Oct-8
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Peptide with sequence C-HMLSHAHQWVTAL corresponding to internal region according to NP_006227.1.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102179-718
- Supplier no.:NBP1-49872
The POU3F3 Antibody from Novus Biologicals is a goat polyclonal antibody to POU3F3. This antibody reacts with mouse, rat. The POU3F3 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: POU3F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: HMLSHAHQWVTAL - internal region
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat