Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor ionotropic N-methyl D-aspartate 1
- Antigen Symbol:NMDAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 μg
- Gene ID:2902
- Antigen Synonyms:glutamate receptor|NMD-R1|ionotropic|NMDAR1glutamate [NMDA] receptor subunit zeta 1|subunit zeta-1|NR1|N-methyl D-aspartate 1|glutamate [NMDA] receptor subunit zeta-1|NMDA1|N-methyl-D-aspartate receptor subunit NR1|N-methyl-D-aspartate receptor channel
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-TQERVNNSNKKE corresponding to internal region according to NP_067544.1, NP_015566.1.
- Tested Applications:, Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102109-002
- Supplier no.:NB100-41105
Specifications
About this item
The NMDAR1 Antibody from Novus Biologicals is a goat polyclonal antibody to NMDAR1. This antibody reacts with rat. The NMDAR1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Peptide ELISA.
Type: Primary
Antigen: NMDAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat