Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 μg
- Storage Conditions:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Gene ID:3028
- Protein Synonyms:MRX31|ERAB3-hydroxyacyl-CoA dehydrogenase type-2|17b-HSD10|EC 1.1.1.35|Mitochondrial RNase P protein 2|hydroxysteroid (17-beta) dehydrogenase 10|syndromic 10|Short-chain type dehydrogenase/reductase XH98G2|MRX17|hydroxyacyl-Coenzyme Adehydrogenase|short chain dehydrogenase/reductase family 5C|short chain L-3-hydroxyacyl-CoA dehydrogenase type 2|amyloid-beta Peptide binding alcohol dehydrogenase|mental retardation|XH98G2|MRPP2HCD2|short chain type dehydrogenase/reductase XH98G2|MRXS10|MHBD|X-linked|type II|17-beta-HSD 10|member 1|SCHAD|HADH2AB-binding alcohol dehydrogenase|CAMR|17-beta-hydroxysteroid dehydrogenase 10|Type II HADH|Mitochondrial ribonuclease P protein 2,3-hydroxyacyl-CoA dehydrogenase type II|ABAD|SDR5C1|DUPXp11.22|hydroxyacyl-Coenzyme A dehydrogenase|3-hydroxy-2-methylbutyryl-CoA dehydrogenase|Endoplasmic reticulum-associated amyloid beta-peptide-binding protein|EC 1.1.1.178
- Protein/Peptide Name:ERAB
- Purity:>95% pure by SDS-PAGE
- Concentration:1 mg/ml
- Tested Applications:ELISA
- Cat. No.:102169-960
- Supplier no.:NBP1-41148
The Recombinant Human ERAB Protein from Novus Biologicals is derived from E. coli. The Recombinant Human ERAB Protein has been validated for the following applications: SDS-Page.