To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 μg
- Storage Conditions:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per 1 microgram of protein (determined by LAL method)
- Enzyme Name:Superoxide Dismutase 1
- Enzyme Synonyms:ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
- Purity:>95% pure by SDS-PAGE
- Concentration:1 mg/ml
- Tested Applications:ELISA
- Cat. No.:102137-422
- Supplier no.:NBC1-18544
The Recombinant Human SOD1/Cu-Zn SOD Protein from Novus Biologicals is derived from E. coli. The Recombinant Human SOD1/Cu-Zn SOD Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
- Entrez Gene ID: 6647
Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.