To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ras-related protein Rab-5
- Antigen Symbol:Rab5a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:5868
- Antigen Synonyms:RAB5RAS-associated protein RAB5A|RAB5A|ras-related protein Rab-5A|member RAS oncogene family
- Storage Buffer:Whole antisera
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:A synthetic peptide from the internal region of mouse Ras-related protein Rab-5A (RAB5A) conjugated to an immunogenic carrier protein has been used as the immunogen. The antigen is homologous in rat.
- Purification:Unpurified
- Cat. No.:102160-072
- Supplier no.:NBP1-20255
Specifications
About this item
The Rab5a Antibody from Novus Biologicals is a sheep polyclonal antibody to Rab5a. This antibody reacts with mouse, rat. The Rab5a Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Rab5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat