Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Toll-like Receptor 4
- Antigen Symbol:TLR4
- Clonality:Monoclonal
- Clone:MTS510
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:100 μg
- Gene ID:7099
- Antigen Synonyms:mts510 tlr4|mts510|toll-like receptor 4|CD284 antigen|CD284|ARMD10|TOLL|hTollhomolog of Drosophila toll
- Storage Buffer:PBS containing 0.05% BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000).
- Purification:Protein G purified
- Cat. No.:102110-112
- Supplier no.:NB100-56560
Specifications
About this item
The TLR4 Antibody (MTS510) from Novus Biologicals is a rat monoclonal antibody to TLR4. This antibody reacts with mouse. The TLR4 Antibody (MTS510) has been validated for the following applications: Flow Cytometry, Immunoprecipitation, Flow (Cell Surface).
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: MTS510
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse