To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Tripartite Motif Containing 47
- Antigen Symbol:TRIM47
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:91107
- Antigen Synonyms:GOARING finger protein 100|tripartite motif containing 47|RNF100tripartite motif-containing protein 47|Gene overexpressed in astrocytoma protein|tripartite motif-containing 47
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 400 and 450 of human tripartite motif-containing 47 (gene over expressed in astrocytoma) using the numbering given in entry NP_258411.2 (GeneID 91107).
- Purification:Immunogen affinity purified
- Cat. No.:102110-836
- Supplier no.:NB100-59842
The TRIM47 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRIM47. This antibody reacts with human. The TRIM47 Antibody has been validated for the following applications: Immunoprecipitation.
Type: Primary
Antigen: TRIM47
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human