About this item
The Rad50 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Rad50. This antibody reacts with human. The Rad50 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunoprecipitation.
Type: Primary
Antigen: Rad50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102105-360
- 102105-362
- Antigen Symbol:
- Rad50
- Rad50
- Antigen Name:
- RAD50 homolog
- RAD50 homolog
- Antigen Synonyms:
- RAD502|RAD50 (S. cerevisiae) homolog|DNA repair protein RAD50|EC 3.6|EC 3.6.1.15|EC 3.6.3.27|RAD50 homolog (S. cerevisiae)|RAD50-2|EC 3.6.3|hRAD50|NBSLD
- RAD502|RAD50 (S. cerevisiae) homolog|DNA repair protein RAD50|EC 3.6|EC 3.6.1.15|EC 3.6.3.27|RAD50 homolog (S. cerevisiae)|RAD50-2|EC 3.6.3|hRAD50|NBSLD
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 10111
- 10111
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- Human RAD50 zinc hook region (amino acids 628-787) expressed in E. coli. [UniProt# Q92878]
- Human RAD50 zinc hook region (amino acids 628-787) expressed in E. coli. [UniProt# Q92878]
- Purification:
- Unpurified
- Unpurified
- Storage Buffer:
- Whole antisera
- Whole antisera
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Tested Applications:
- Simple Western,
- Simple Western,