To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl CPG Binding Protein 2
- Antigen Symbol:MeCP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50ug
- Environmentally Preferable:
- Gene ID:4204
- Antigen Synonyms:methyl CpG binding protein 2 (Rett syndrome)|X-linked 79|mental retardation|DKFZp686A24160|AUTSX3|MeCp-2 protein|X-linked 16
- Storage Buffer:PBS with 1 mg/ml BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic Peptide: M(1) V A G M L G L R E E K S E D(15) C
- Purification:Immunogen affinity purified
- Cat. No.:102132-962
- Supplier no.:NB600-1101
Specifications
About this item
Rabbit Polyclonal MeCP2 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat.
- There is 1 scientific publication using this antibody.
Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human