To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Microtubule-associated protein 1A
- Antigen Symbol:MAP1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4130
- Antigen Synonyms:MAP-1A|Proliferation-related protein p80|FLJ77111|MAP1LMTAP1A|EC 6.3.4.5|EC 2.7.2.8|microtubule-associated protein 1A
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human microtubule-associated protein 1A using the numbering given in entry NP_002364.5 (GeneID 4130).
- Purification:Immunogen affinity purified
- Cat. No.:102115-596
- Supplier no.:NB100-68254
Specifications
About this item
Rabbit Polyclonal MAP1A Antibody. Tested Applications: Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: MAP1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human