Rabbit Polyclonal LIMPII/lpg85 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat, Canine.
Type: Primary
Antigen: LIMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 102131-474
- 102131-472
- Antigen Symbol:
- LIMPII/lpg85
- LIMPII/lpg85
- Antigen Name:
- Lysosome membrane protein 2
- Lysosome membrane protein 2
- Antigen Synonyms:
- thrombospondin receptor)-like 2(lysosomal integral membrane protein II)|LIMPIICD36 antigen-like 2|lysosome membrane protein 2|AMRF|Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein|LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B|Lysosome membrane protein II|scavenger receptor class B|member 2|SR-BII|LGP85|CD36 antigen|CD36 antigen (collagen type I receptor|CD36L2LIMP II|HLGP85
- thrombospondin receptor)-like 2(lysosomal integral membrane protein II)|LIMPIICD36 antigen-like 2|lysosome membrane protein 2|AMRF|Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein|LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B|Lysosome membrane protein II|scavenger receptor class B|member 2|SR-BII|LGP85|CD36 antigen|CD36 antigen (collagen type I receptor|CD36L2LIMP II|HLGP85
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Primates, Human, Dog, Rat, Mouse
- Primates, Human, Dog, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 950
- 950
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A C-terminal synthetic peptide made to the mouse LIMPII/lgp85 protein sequence. [UniProt# O35114]
- A C-terminal synthetic peptide made to the mouse LIMPII/lgp85 protein sequence. [UniProt# O35114]
- Purification:
- Unpurified
- Unpurified
- Storage Buffer:
- Whole antisera
- Whole antisera
- Storage Temperature:
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.