Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:gamma H2AX
- Antigen Symbol:H2AXG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:3014
- Antigen Synonyms:member X|H2AX histone|H2a/x|H2A.X|H2AXhistone H2A.x|H2A histone family|YH2AX
- Modification:p Ser139
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:Immunogen was a synthetic phospho-peptide, which represented a portion of the C-terminus of human histone H2AX surrounding phosphorylated serine 139 (GeneID 3014).
- Purification:Immunogen affinity purified
- Cat. No.:102106-982
- Supplier no.:NB100-2280
Specifications
About this item
The gamma H2AX [p Ser139] Antibody from Novus Biologicals is a rabbit polyclonal antibody to gamma H2AX. This antibody reacts with human, mouse. The gamma H2AX [p Ser139] Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500
Type: Primary
Antigen: gamma H2AX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse