Order Entry
Puerto Rico
ContactUsLinkComponent
ND OLLAS/DYKDDDDK Vector, Novus Biologicals
ND OLLAS/DYKDDDDK Vector, Novus Biologicals
Catalog # 102136-984
Supplier:  Novus Biologicals
CAS Number:  
undefined
ND OLLAS/DYKDDDDK Vector, Novus Biologicals
Catalog # 102136-984
Supplier:  Novus Biologicals
Supplier Number:  NBA1-07084
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    10 µg
  • Cat. No.:
    102136-984
  • Supplier no.:
    NBA1-07084

Specifications

About this item

The pCMV-N-DYK-OLLAS vector provides a convenient vector to express recombinant proteins under control of the CMV promoter with both an N-terminal OLLAS tag and a DYKDDDDK tag. Using this vector the protein will be expressed in the cytoplasm. We recommend inserting your protein of interest into the vector in frame using the Xba I and EcoR I/Not I restriction sites shown above. Bacterial selection can be performed using Kanamycin, while Neomycin resistance can be used for screening mammalian transfectants. The OLLAS tag has been shown to be detectable at ~100-fold lower levels that similar commercially available tags using the OLLA-2 monoclonal antibody. The OLLAS tag has also been demonstrated to provide superior functionality whether used as an N-terminal, C-terminal, or internal tag on recombinant and fusion proteins without interfering with the biological activity of the tagged protein.

  • Buffer: 5 mM Tris pH 8.0
  • Specificity: pCMV-ND OLLAS DYKDDDDK Vector for the intracellular expression of N-terminally FLAG(DYKDDDDK) and OLLAS dual tandem tagged protein.
  • Tested applications: Western Blot

Application Notes: Using this vector the protein will be expressed in the cytoplasm. We recommend inserting your protein of interest into the vector in frame using the Xba I or EcoRI/Not I restriction sites shown above. Bacterial selection can be performed using Kanamycin, while Neomycin resistance can be used for screening mammalian transfectants.

Alternate Names for ND OLLAS/DYKDDDDK Vector:
DDDDK vector
DYKDDDDK epitope tag vector
DYKDDDDK vector
ECS tag vector
ECS vector
Enterokinase Cleavage Site epitope tag vector
Enterokinase Cleavage Site tag vector
FLAG vector (FLAG is a trademark of Sigma Aldrich)
OLLA-2 vector
xxxDDDDK epitope tag vector

Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.