Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Plexin Domain Containing 1
- Antigen Symbol:TEM7/PLXDC1
- Clonality:Monoclonal
- Clone:197C193 (IM193)
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:57125
- Antigen Synonyms:Tumor endothelial marker 7|TEM7DKFZp686F0937|plexin domain containing 1,2410003I07Rik|TEM3FLJ36270|FLJ45632|Tumor endothelial marker 3|plexin domain-containing protein 1
- Storage Buffer:PBS containing 0.05% BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Amino acids 409-425 (LQNNLSPKTKGTPVHLG) of human TEM7 were used to develop this monoclonal antibody.
- Purification:Protein G purified
- Cat. No.:102110-108
- Supplier no.:NB100-56557
The TEM7 / PLXDC1 Antibody (197C193 (IM193)) from Novus Biologicals is a mouse monoclonal antibody to TEM7 / PLXDC1. This antibody reacts with human, mouse, rat. The TEM7 / PLXDC1 Antibody (197C193 (IM193)) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: TEM7/PLXDC1
Clonality: Monoclonal
Clone: 197C193 (IM193)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat