Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:RAD51 Paralog C
- Antigen Symbol:Rad51C
- Clonality:Monoclonal
- Clone:2H11/6
- Conjugation:DyLight® 488
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Yeast,Primates,Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:5889
- Antigen Synonyms:yeast RAD51 homolog 3|DNA repair protein RAD51 homolog 3|MGC104277|RAD51 homolog C|RAD51L2RAD51 (S. cerevisiae) homolog C|BROVCA3|RAD51 homolog C (S. cerevisiae)|FANCO|R51H3|RAD51-like protein 2
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:His-tagged human Rad51C, over-expressed in E. coli. [UniProt# O43502]
- Purification:Protein G purified
- Cat. No.:102105-928
- Supplier no.:NB100-177G
Specifications
About this item
The Rad51C Antibody (2H11 / 6) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to Rad51C. This antibody reacts with human, mouse, primate, yeast. The Rad51C Antibody (2H11 / 6) [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Rad51C
Clonality: Monoclonal
Clone: 2H11/6
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Primate, Yeast