Order Entry
United States
ContactUsLinkComponent
Anti-OATP1B1/OATP2 Mouse Monoclonal Antibody [clone: ESL]
Anti-OATP1B1/OATP2 Mouse Monoclonal Antibody [clone: ESL]
Catalog # 102116-402
Supplier:  Novus Biologicals
Anti-OATP1B1/OATP2 Mouse Monoclonal Antibody [clone: ESL]
Catalog # 102116-402
Supplier:  Novus Biologicals
Supplier Number:  NB100-74481
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Organic anion transporter polypeptide 2
  • Antigen Symbol:
    OATP1B1/OATP2
  • Clonality:
    Monoclonal
  • Clone:
    ESL
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100μl
  • Environmentally Preferable:
  • Gene ID:
    10599
  • Antigen Synonyms:
    OATP-COATP-2|member 6|member 1B1|solute carrier organic anion transporter family|LST-1Solute carrier family 21 member 6|OATP2MGC133282|OATPCsolute carrier organic anion transporter family member 1B1|OATP1B1Sodium-independent organic anion-transporting polypeptide 2|solute carrier family 21 (organic anion transporter)|LST1SLC21A6|Liver-specific organic anion transporter 1
  • Storage Buffer:
    Tissue culture supernatant
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide corresponding to residues E(671) S L N K N K H F V P S A G A D S E T H C(691) of human OATP2.
  • Purification:
    Unpurified
  • Cat. No.:
    102116-402
  • Supplier no.:
    NB100-74481

Specifications

About this item

The OATP1B1 / OATP2 Antibody (ESL) from Novus Biologicals is a mouse monoclonal antibody to OATP1B1 / OATP2. This antibody reacts with human, canine (negative), mouse (negative), rat (negative). The OATP1B1 / OATP2 Antibody (ESL) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: OATP1B1/OATP2
Clonality: Monoclonal
Clone: ESL
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Canine (Negative), Mouse (Negative), Rat (Negative)