To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Huntingtin
- Antigen Symbol:Huntingtin
- Clonality:Monoclonal
- Clone:HDB4E10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:3064
- Antigen Synonyms:HD protein|IT15HDHuntington disease protein|huntingtin|huntingtin (Huntington disease)
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Recombinant fragment, corresponding to amino acids 1844-2131 of Huntingtin.
- Purification:Protein A or G purified
- Cat. No.:102133-060
- Supplier no.:NB600-1197
Specifications
About this item
Mouse Monoclonal Huntingtin Antibody (HDB4E10). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: HTT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human