To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Fibroblast Surface Protein
- Antigen Symbol:Fibroblast Surface Protein
- Clonality:Monoclonal
- Clone:1B10
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Human
- Size:100 μl
- Storage Buffer:The product is provided as the immunoglobulin fraction of mouse ascites fluid in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 0.1% sodium azide as a preservative.
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.2 mg/ml
- Immunogen:Fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with cultured human thymic fibroblasts.
- Purification:Unpurified
- Cat. No.:102106-192
- Supplier no.:NB100-1845
Specifications
About this item
The Fibroblast Surface Protein Antibody (1B10) from Novus Biologicals is a mouse monoclonal antibody to Fibroblast Surface Protein. This antibody reacts with human. The Fibroblast Surface Protein Antibody (1B10) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Type: Primary
Antigen: Fibroblast Surface Protein
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human