To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Integrin alpha 2b
- Antigen Symbol:ITGA2b
- Clonality:Monoclonal
- Clone:PM6/248
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Baboon,Mustelid,Human
- Size:100 μg
- Gene ID:3674
- Antigen Synonyms:alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex|GTA|platelet fibrinogen receptor|GP2Bintegrin alpha-IIb|alpha subunit|antigenCD41)|GPIIb|CD41|Platelet membrane glycoprotein IIb|antigenCD41B)|CD41 antigen|ITGAB|CD41BHPA3|integrin|platelet-specific antigen BAK|GPalpha IIb
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.1 mg/ml
- Immunogen:Human platelet plasma membrane
- Purification:Protein G purified
- Cat. No.:102112-998
- Supplier no.:NB100-63782
Mouse Monoclonal CD41/Integrin alpha 2 beta Antibody (PM6/248) [FITC]. Tested Applications: Flow Cytometry. Tested Reactivity: Human.
Type: Primary
Antigen: CD41/Integrin alpha 2 beta
Clonality: Monoclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: