To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:160 kDa Neurofilament Medium
- Antigen Symbol:NF-M
- Clonality:Monoclonal
- Clone:3H11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Birds,Mammals,Human,Rat,Mouse
- Western Blot:Yes
- Size:0.05 ml
- Environmentally Preferable:
- Gene ID:4741
- Antigen Synonyms:Neurofilament 3|medium polypeptide 150kDa|NEF3neurofilament medium polypeptide|neurofilament|NFMNeurofilament triplet M protein|neurofilament-3 (150 kD medium)|medium polypeptide|NF-M160 kDa neurofilament protein
- Storage Buffer:Ascites
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Recombinant rat Neurofilament Medium fusion protein corresponding to the C-terminus [UniProt# P12839]
- Purification:Unpurified
- Cat. No.:102129-982
- Supplier no.:NB300-134
The NF-M Antibody (3H11) from Novus Biologicals is a mouse monoclonal antibody to NF-M. This antibody reacts with human, mouse, rat, avian, mammal. The NF-M Antibody (3H11) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:500
Type: Primary
Antigen: NF-M
Clonality: Monoclonal
Clone: 3H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Avian, Mammal