Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Pentraxin 3/TSG-14 (from E. coli)
Human Recombinant Pentraxin 3/TSG-14 (from E. coli)
Catalog # 102137-636
Supplier:  Novus Biologicals
CAS Number:  
Human Recombinant Pentraxin 3/TSG-14 (from E. coli)
Catalog # 102137-636
Supplier:  Novus Biologicals
Supplier Number:  NBC1-26376
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 μg
  • Storage Conditions:
    Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
  • Gene ID:
    5806
  • Protein Synonyms:
    long|tumor necrosis factor|pentraxin 3|pentraxin-related protein PTX3|TSG-14pentaxin-related gene|pentaxin-related gene|TNF alpha-induced protein 5|Pentaxin-related protein PTX3|TNFAIP5|tumor necrosis factor-inducible protein TSG-14|TSG14|alpha-induced protein 5|pentraxin-3|pentraxin-related gene|Tumor necrosis factor alpha-induced protein 5|Tumor necrosis factor-inducible gene 14 protein|rapidly induced by IL-1 beta
  • Protein/Peptide Name:
    Pentraxin 3/TSG-14
  • Purity:
    >90%, by SDS-PAGE
  • Concentration:
    1 mg/ml
  • Tested Applications:
    ELISA
  • Cat. No.:
    102137-636
  • Supplier no.:
    NBC1-26376

Specifications

About this item

The Recombinant Human Pentraxin 3/TSG-14 Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Pentraxin 3/TSG-14 Protein has been validated for the following applications: SDS-Page.