To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 μg
- Storage Conditions:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Enzyme Name:Hexokinase Type III
- Enzyme Synonyms:ATP:D-hexose 6-phosphotransferase, EC 2.7.1, EC 2.7.1.1, hexokinase 3 (white cell), Hexokinase type III, hexokinase-3, HK III, HKIII, HXK3
- Purity:>95% pure by SDS-PAGE
- Concentration:1 mg/ml
- Tested Applications:ELISA
- Cat. No.:102137-132
- Supplier no.:NBC1-18387
The Recombinant Human Hexokinase Type III Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Hexokinase Type III Protein has been validated for the following applications: SDS-Page.
- Entrez Gene ID: 3101
Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.