Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Amine Oxidase, Copper Containing 3
- Antigen Symbol:VAP-1/AOC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Epitope:DEDPSFYSADSIY
- Gene ID:8639
- Antigen Synonyms:Vascular adhesion protein 1|EC 1.4.3|Semicarbazide-sensitive amine oxidase|VAP-1membrane primary amine oxidase|copper containing 3 (vascular adhesion protein 1)|HPAOSSAO|VAP1EC 1.4.3.21|Copper amine oxidase|amine oxidase
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-DEDPSFYSADSIY corresponding to internal region (near C-Terminus) according to NP_003725.1.
- Tested Applications:, Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102110-342
- Supplier no.:NB100-57090
The VAP-1 / AOC3 Antibody from Novus Biologicals is a goat polyclonal antibody to VAP-1 / AOC3. This antibody reacts with human. The VAP-1 / AOC3 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: VAP-1/AOC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: DEDPSFYSADSIY
Host: Goat
Isotype: IgG
Reactivity: Human