Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Dyskeratosis Congenita 1, Dyskerin
- Antigen Symbol:DKC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Epitope:KRKRESESESDETPP
- Gene ID:1736
- Antigen Synonyms:Dyskerin|DKC|snoRNP protein DKC1|Nucleolar protein family A member 4|Nopp140-associated protein of 57 kDa|dyskerin|CBF5|dyskeratosis congenita 1|EC 5.4.99.-|cbf5p homolog|XAP101|CBF5 homolog|FLJ97620|EC 5.4.99|H/ACA ribonucleoprotein complex subunit 4|NOLA4dyskerin|NAP57|Nucleolar protein NAP57
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-KRKRESESESDETPP corresponding to internal region according to NP_001354.1.
- Purification:Immunogen affinity purified
- Cat. No.:102110-742
- Supplier no.:NB100-59738
Specifications
About this item
The DKC1 Antibody from Novus Biologicals is a goat polyclonal antibody to DKC1. This antibody reacts with human. The DKC1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: DKC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: KRKRESESESDETPP
Host: Goat
Isotype: IgG
Reactivity: Human