Order Entry
Puerto Rico
ContactUsLinkComponent
Ammonia solution 29%, Cleanroom® MB for the electronics industry
Corrosive
Irritant
Environment
Ammonia solution 29%, Cleanroom® MB for the electronics industry
Catalog # 71009-320
Supplier:  KMG
CAS Number:  
Corrosive
Irritant
Environment
Ammonia solution 29%, Cleanroom® MB for the electronics industry
Catalog # 71009-320
Supplier:  KMG
Supplier Number:  63925
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle
  • Size:
    1 gal.
  • Cat. No.:
    71009-320
  • Supplier no.:
    405-063925

Specifications

Specification Test Results

  • Assay (NH3):
    28.00 to 30.00 %
  • Color:
    10 APHA
  • Appearance:
    Conforms
  • Substance Reducing KMnO4:
    Conforms
  • Carbon Dioxide (CO2):
    10,000 ppb
  • Chloride (Cl):
    200 ppb
  • Nitrate (NO3):
    400 ppb
  • Phosphate (PO4):
    100 ppb
  • Sulfate (SO4):
    200 ppb
  • Aluminum (Al):
    10.0 ppb
  • Antimony (Sb):
    5.0 ppb
  • Arsenic (As):
    5.0 ppb
  • Barium (Ba):
    10.0 ppb
  • Beryllium (Be):
    10.0 ppb
  • Bismuth (Bi):
    10.0 ppb
  • Boron (B):
    10.0 ppb
  • Cadmium (Cd):
    10.0 ppb
  • Calcium (Ca):
    10.0 ppb
  • Chromium (Cr):
    10.0 ppb
  • Cobalt (Co):
    10.0 ppb
  • Copper (Cu):
    10.0 ppb
  • Gallium (Ga):
    10.0 ppb
  • Germanium (Ge):
    10.0 ppb
  • Gold (Au):
    10.0 ppb
  • Iron (Fe):
    10.0 ppb
  • Lead (Pb):
    10.0 ppb
  • Lithium (Li):
    10.0 ppb
  • Magnesium (Mg):
    10.0 ppb
  • Manganese (Mn):
    10.0 ppb
  • Molybdenum (Mo):
    10.0 ppb
  • Nickel (Ni):
    10.0 ppb
  • Niobium (Nb):
    10.0 ppb
  • Potassium (K):
    10.0 ppb
  • Silicon (Si):
    10.0 ppb
  • Silver (Ag):
    10.0 ppb
  • Sodium (Na):
    10.0 ppb
  • Strontium (Sr):
    10.0 ppb
  • Tantalum (Ta):
    10.0 ppb
  • Thallium (Tl):
    10.0 ppb
  • Tin (Sn):
    10.0 ppb
  • Titanium (Ti):
    10.0 ppb
  • Vanadium (V):
    10.0 ppb
  • Zinc (Zn):
    10.0 ppb
  • Zirconium (Zr):
    10.0 ppb
  • 0.5μ Particle Count:
    25 par/ml
  • 1.0μ Particle Count:
    10 par/ml

Specification Test Results

About this item

 :
 :35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650