About this item
The E.Z.N.A.® Viral RNA Kit is designed for the isolation of viral RNA from cell-free fluids such as plasma, serum, urine, and cell culture supernatant
The procedure completely removes contaminants and enzyme inhibitors making viral RNA isolation fast, convenient, and reliable. The kit is also suitable for the isolation of total RNA from cultured cells, tissues, and bacteria. RNA purified using the E.Z.N.A.® Viral RNA method is ready for all downstream applications such as RT-PCR.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: