Corning® 3M™ Sodium Acetate, Corning
Catalog # 45001-124
Supplier: Corning
CAS Number:
Specifications
- Description:3M Sodium Acetate
- Size:100 mL
- Storage Temperature:Ambient
- Shipping Temperature:Ambient
- Cat. No.:45001-124
- Supplier no.:46-033-CI
Specifications
About this item
Sodium Acetate is a common molecular biology reagent. All production lots are tested to ensure that they are free of RNAse. It is often used in buffer systems to maintain the pH of a given solution.